Assay Details
| Gene Symbol: ISL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018065772.1 | XP_017921261.1 | 5 - 6 | 1459 | 57 | |
| GenBank mRNA | JO364509.2 | - | 1 - 2 | 104 | 57 | |
More Information
Set Membership: |
|

ISL1
Genomic Map