Assay Details
| Gene Symbol: CLDN18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005675525.3 | XP_005675582.1 | 2 - 3 | 517 | 62 | |
| XM_005675526.3 | XP_005675583.2 | 2 - 3 | 514 | 62 | ||
More Information
Set Membership: |
|

CLDN18
Genomic Map