Assay Details
| Gene Symbol: PRELID2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005683091.3 | XP_005683148.1 | 4 - 5 | 587 | 78 | |
| GenBank mRNA | JO602005.2 | - | 4 - 5 | 485 | 78 | |
More Information
Set Membership: |
|

PRELID2
Genomic Map