Assay Details
| Gene Symbol: TMX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005685645.3 | XP_005685702.1 | 6 - 7 | 845 | 74 | |
| GenBank mRNA | JO436508.2 | - | 2 - 3 | 584 | 74 | |
More Information
Set Membership: |
|

TMX1
Genomic Map