Assay Details
| Gene Symbol: NYAP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018041020.1 | XP_017896509.1 | 5 - 6 | 2355 | 87 | |
| XM_018041021.1 | XP_017896510.1 | 5 - 6 | 2412 | 87 | ||
| XM_018041022.1 | XP_017896511.1 | 4 - 5 | 2163 | 87 | ||
| GenBank mRNA | JO643781.2 | - | 2 - 3 | 1144 | 87 | |
More Information
Set Membership: |
|

NYAP1
Genomic Map