Assay Details
| Gene Symbol: ANGPTL8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005682377.3 | XP_005682434.1 | 1 - 2 | 385 | 62 | |
| GenBank mRNA | JO411045.2 | - | 1 - 2 | 350 | 62 | |
More Information
Set Membership: |
|

ANGPTL8
Genomic Map