Assay Details
| Gene Symbol: GLRA3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018052791.1 | XP_017908280.1 | 7 - 8 | 940 | 82 | |
| XM_018052792.1 | XP_017908281.1 | 7 - 8 | 925 | 82 | ||
More Information
Set Membership: |
|

GLRA3
Genomic Map