Assay Details
Gene Symbol: HBEGF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018050289.1 | XP_017905778.1 | 4 - 5 | 1226 | 56 | |
XM_018050290.1 | XP_017905779.1 | 4 - 5 | 1623 | 56 | ||
GenBank mRNA | KA336377.1 | - | 3 - 4 | 706 | 56 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
