Assay Details
| Gene Symbol: MARCH8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018042357.1 | XP_017897846.1 | 6 - 7 | 783 | 64 | |
| XM_018042358.1 | XP_017897847.1 | 7 - 8 | 1629 | 64 | ||
| GenBank mRNA | JO325820.2 | - | 6 - 7 | 704 | 64 | |
More Information
Set Membership: |
|

MARCH8
Genomic Map