Assay Details
| Gene Symbol: WFDC5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005688586.3 | XP_005688643.1 | 3 - 4 | 451 | 65 | |
| GenBank mRNA | KA326283.1 | - | 3 - 4 | 380 | 65 | |
More Information
Set Membership: |
|

WFDC5
Genomic Map