Assay Details
| Gene Symbol: SLC22A14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018038485.1 | XP_017893974.1 | 7 - 8 | 1374 | 63 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SLC22A14| Gene Symbol: SLC22A14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018038485.1 | XP_017893974.1 | 7 - 8 | 1374 | 63 | |
Set Membership: |
|