Assay Details
| Gene Symbol: LOC102173702 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005696422.3 | XP_005696479.2 | 5 - 6 | 583 | 97 | |
| GenBank mRNA | JO421205.2 | - | 2 - 3 | 569 | 97 | |
| JO439462.2 | - | 5 - 6 | 656 | 97 | ||
More Information
Set Membership: |
|

LOC102173702
Genomic Map