Assay Details
| Gene Symbol: SERPINB7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005697316.2 | XP_005697373.2 | 6 - 7 | 678 | 59 | |
| XM_005697317.2 | XP_005697374.2 | 6 - 7 | 675 | 59 | ||
| XM_018039813.1 | XP_017895302.1 | 6 - 7 | 633 | 59 | ||
| GenBank mRNA | KA346511.1 | - | 2 - 3 | 1285 | 59 | |
More Information
Set Membership: |
|

SERPINB7
Genomic Map