Assay Details
| Gene Symbol: FAM221A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018047064.1 | XP_017902553.1 | 3 - 4 | 486 | 70 | |
| XM_018047065.1 | XP_017902554.1 | 3 - 4 | 486 | 70 | ||
| XM_018047066.1 | XP_017902555.1 | 3 - 4 | 486 | 70 | ||
| GenBank mRNA | JO442065.2 | - | 4 - 5 | 511 | 70 | |
More Information
Set Membership: |
|

FAM221A
Genomic Map