Assay Details
| Gene Symbol: SNAPC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018050991.1 | XP_017906480.1 | 3 - 4 | 437 | 59 | |
| XM_018050992.1 | XP_017906481.1 | 3 - 4 | 609 | 59 | ||
| GenBank mRNA | JO629970.2 | - | 3 - 4 | 431 | 59 | |
| JO629971.2 | - | 3 - 4 | 431 | 59 | ||
| KA307346.1 | - | 3 - 4 | 541 | 59 | ||
More Information
Set Membership: |
|

SNAPC2
Genomic Map