Assay Details
Gene Symbol: PTS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018059782.1 | XP_017915271.1 | 2 - 3 | 251 | 61 | |
GenBank mRNA | JO605637.2 | - | 4 - 5 | 513 | 61 | |
JT644783.1 | - | 2 - 3 | 120 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
