Assay Details
| Gene Symbol: EXT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018059215.1 | XP_017914704.1 | 13 - 14 | 2516 | 64 | |
| GenBank mRNA | JO496455.2 | - | 13 - 14 | 2274 | 64 | |
| KA310482.1 | - | 1 - 2 | 691 | 64 | ||
More Information
Set Membership: |
|

EXT2
Genomic Map