Assay Details
| Gene Symbol: ZNF527 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005692364.3 | XP_005692421.1 | 4 - 5 | 1194 | 88 | |
| XM_013971160.2 | XP_013826614.1 | 4 - 5 | 1318 | 88 | ||
| GenBank mRNA | KA311967.1 | - | 2 - 3 | 310 | 88 | |
More Information
Set Membership: |
|

ZNF527
Genomic Map