Assay Details
| Gene Symbol: ZNF527 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005692364.3 | XP_005692421.1 | 5 - 6 | 1289 | 76 | |
| XM_005692365.3 | XP_005692422.1 | 5 - 6 | 1279 | 76 | ||
| XM_013971160.2 | XP_013826614.1 | 5 - 6 | 1413 | 76 | ||
| GenBank mRNA | JT678663.1 | - | 4 - 5 | 346 | 76 | |
| KA311967.1 | - | 1 - 2 | 215 | 76 | ||
More Information
Set Membership: |
|

ZNF527
Genomic Map