Assay Details
| Gene Symbol: ZNF432 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005692878.3 | XP_005692935.2 | 4 - 5 | 556 | 68 | |
| GenBank mRNA | JO626610.2 | - | 1 - 2 | 154 | 68 | |
More Information
Set Membership: |
|

ZNF432
Genomic Map