Assay Details
| Gene Symbol: ZGRF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013964465.2 | XP_013819919.2 | 9 - 10 | 3119 | 74 | |
| XM_018049209.1 | XP_017904698.1 | 9 - 10 | 3119 | 74 | ||
| XM_018049210.1 | XP_017904699.1 | 9 - 10 | 3119 | 74 | ||
| XM_018049211.1 | XP_017904700.1 | 9 - 10 | 3119 | 74 | ||
| XM_018049212.1 | XP_017904701.1 | 9 - 10 | 1646 | 74 | ||
More Information
Set Membership: |
|

ZGRF1
Genomic Map