Assay Details
| Gene Symbol: FAM83B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018039254.1 | XP_017894743.1 | 4 - 5 | 903 | 78 | |
| GenBank mRNA | KA328079.1 | - | 1 - 2 | 2339 | 78 | |
More Information
Set Membership: |
|

FAM83B
Genomic Map