Assay Details
| Gene Symbol: FAM192A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005692012.3 | XP_005692069.2 | 6 - 7 | 606 | 61 | |
| XM_005692015.3 | XP_005692072.2 | 6 - 7 | 580 | 61 | ||
| XM_018062034.1 | XP_017917523.1 | 6 - 7 | 770 | 61 | ||
| XM_018062035.1 | XP_017917524.1 | 7 - 8 | 890 | 61 | ||
| XM_018062036.1 | XP_017917525.1 | 6 - 7 | 766 | 61 | ||
| GenBank mRNA | JO434062.2 | - | 1 - 2 | 825 | 61 | |
| KA346341.1 | - | 6 - 7 | 747 | 61 | ||
More Information
Set Membership: |
|

FAM192A
Genomic Map