Assay Details
| Gene Symbol: GLYATL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005696397.3 | XP_005696454.2 | 1 - 2 | 89 | 92 | |
| GenBank mRNA | JO397937.2 | - | 1 - 2 | 95 | 92 | |
More Information
Set Membership: |
|

GLYATL3
Genomic Map