Assay Details
| Gene Symbol: ZNF652 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018064915.1 | XP_017920404.1 | 5 - 6 | 1804 | 72 | |
| XM_018064916.1 | XP_017920405.1 | 4 - 5 | 1635 | 72 | ||
| XM_018064917.1 | XP_017920406.1 | 4 - 5 | 1707 | 72 | ||
| XM_018064918.1 | XP_017920407.1 | 4 - 5 | 2644 | 72 | ||
More Information
Set Membership: |
|

ZNF652
Genomic Map