Assay Details
| Gene Symbol: CDK15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005676377.3 | XP_005676434.2 | 6 - 7 | 662 | 61 | |
| XM_005676378.3 | XP_005676435.2 | 6 - 7 | 662 | 61 | ||
More Information
Set Membership: |
|

CDK15
Genomic Map