Assay Details
| Gene Symbol: ANKRD33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018047741.1 | XP_017903230.1 | 3 - 4 | 425 | 65 | |
| XM_018047742.1 | XP_017903231.1 | 3 - 4 | 341 | 65 | ||
| GenBank mRNA | KA330516.1 | - | 1 - 2 | 236 | 65 | |
More Information
Set Membership: |
|

ANKRD33
Genomic Map