Assay Details
| Gene Symbol: LAYN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018059803.1 | XP_017915292.1 | 6 - 7 | 895 | 74 | |
| XM_018059804.1 | XP_017915293.1 | 5 - 6 | 879 | 74 | ||
| XM_018059805.1 | XP_017915294.1 | 6 - 7 | 828 | 74 | ||
| XM_018059806.1 | XP_017915295.1 | 5 - 6 | 447 | 74 | ||
| GenBank mRNA | JT707895.1 | - | 6 - 7 | 918 | 74 | |
More Information
Set Membership: |
|

LAYN
Genomic Map