Assay Details
| Gene Symbol: GLTP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013970784.2 | XP_013826238.2 | 2 - 3 | 312 | 65 | |
| XM_018061107.1 | XP_017916596.1 | 2 - 3 | 130 | 65 | ||
| GenBank mRNA | KA312862.1 | - | 2 - 3 | 317 | 65 | |
More Information
Set Membership: |
|

GLTP
Genomic Map