Assay Details
| Gene Symbol: SOX30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018051717.1 | XP_017907206.1 | 5 - 6 | 1441 | 64 | |
| XM_018051718.1 | XP_017907207.1 | 3 - 4 | 1734 | 64 | ||
More Information
Set Membership: |
|

SOX30
Genomic Map