Assay Details
| Gene Symbol: MCMDC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005689035.3 | XP_005689092.1 | 12 - 13 | 1775 | 74 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

MCMDC2| Gene Symbol: MCMDC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005689035.3 | XP_005689092.1 | 12 - 13 | 1775 | 74 | |
Set Membership: |
|