Assay Details
| Gene Symbol: KLK12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005692755.3 | XP_005692812.2 | 4 - 5 | 590 | 102 | |
| XM_005692756.3 | XP_005692813.1 | 4 - 5 | 582 | 102 | ||
| XM_005692757.3 | XP_005692814.1 | 4 - 5 | 675 | 102 | ||
| GenBank mRNA | JO369246.2 | - | 2 - 3 | 495 | 102 | |
| JO512088.2 | - | 2 - 3 | 307 | 102 | ||
More Information
Set Membership: |
|

KLK12
Genomic Map