Assay Details
| Gene Symbol: ZNF711 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005700578.2 | XP_005700635.1 | 4 - 5 | 934 | 76 | |
| XM_013976375.2 | XP_013831829.1 | 4 - 5 | 1743 | 76 | ||
| XM_013976376.2 | XP_013831830.1 | 4 - 5 | 1876 | 76 | ||
| XM_013976377.2 | XP_013831831.1 | 4 - 5 | 1708 | 76 | ||
| XM_013976378.2 | XP_013831832.1 | 3 - 4 | 1140 | 76 | ||
More Information
Set Membership: |
|

ZNF711
Genomic Map