Assay Details
Gene Symbol: JPT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018040035.1 | XP_017895524.1 | 3 - 4 | 392 | 106 | |
GenBank mRNA | JO426325.1 | - | 3 - 4 | 437 | 106 | |
JO428335.1 | - | 3 - 4 | 437 | 106 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
