Assay Details
| Gene Symbol: CLDN15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013974836.2 | XP_013830290.1 | 3 - 4 | 2050 | 70 | |
| XM_018040877.1 | XP_017896366.1 | 4 - 5 | 1910 | 70 | ||
| XM_018040878.1 | XP_017896367.1 | 4 - 5 | 1933 | 70 | ||
| XM_018040879.1 | XP_017896368.1 | 3 - 4 | 1984 | 70 | ||
| XM_018040880.1 | XP_017896369.1 | 3 - 4 | 2277 | 70 | ||
More Information
Set Membership: |
|

CLDN15
Genomic Map