Assay Details
| Gene Symbol: ZDHHC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018067002.1 | XP_017922491.1 | 5 - 6 | 885 | 65 | |
| XM_018067003.1 | XP_017922492.1 | 5 - 6 | 894 | 65 | ||
| XM_018067005.1 | XP_017922494.1 | 5 - 6 | 885 | 65 | ||
| GenBank mRNA | JT750762.1 | - | 5 - 6 | 846 | 65 | |
More Information
Set Membership: |
|

ZDHHC3
Genomic Map