Assay Details
Gene Symbol: LGI4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018063349.1 | XP_017918838.1 | 7 - 8 | 1142 | 89 | |
XM_018063351.1 | XP_017918840.1 | 5 - 6 | 967 | 89 | ||
XM_018063352.1 | XP_017918841.1 | 7 - 8 | 1158 | 89 | ||
XM_018063353.1 | XP_017918842.1 | 7 - 8 | 1154 | 89 | ||
XM_018063354.1 | XP_017918843.1 | 7 - 8 | 1178 | 89 | ||
XM_018063355.1 | XP_017918844.1 | 7 - 8 | 1087 | 89 | ||
XM_018063356.1 | XP_017918845.1 | 7 - 8 | 1083 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
