Assay Details
| Gene Symbol: RNF26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005689543.3 | XP_005689600.1 | 1 | 1238 | 101 | |
| GenBank mRNA | JO577235.1 | - | 1 | 1282 | 101 | |
| JO621169.2 | - | 1 | 1232 | 101 | ||
More Information
Set Membership: |
|

RNF26
Genomic Map