Assay Details
Gene Symbol: RGS8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005690949.3 | XP_005691006.1 | 3 - 4 | 636 | 65 | |
XM_013970422.2 | XP_013825876.1 | 4 - 5 | 430 | 65 | ||
XM_013970423.2 | XP_013825877.1 | 4 - 5 | 361 | 65 | ||
XM_013970424.2 | XP_013825878.1 | 4 - 5 | 498 | 65 | ||
XM_018060690.1 | XP_017916179.1 | 6 - 7 | 667 | 65 | ||
GenBank mRNA | JO644458.2 | - | 6 - 7 | 778 | 65 | |
JO644459.2 | - | 6 - 7 | 778 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
