Assay Details
Gene Symbol: NDUFB4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005674988.3 | XP_005675045.2 | 1 - 2 | 311 | 74 | |
GenBank mRNA | JO375330.2 | - | 1 - 2 | 244 | 74 | |
JO557335.2 | - | 278 | 74 | |||
JO557336.2 | - | 1 - 2 | 263 | 74 | ||
JO612247.2 | - | 1 - 2 | 318 | 74 | ||
JO644795.2 | - | 1 - 2 | 405 | 74 | ||
JR125738.1 | - | 1 - 2 | 128 | 74 | ||
JT673041.1 | - | 1 - 2 | 154 | 74 | ||
JT674050.1 | - | 2 - 3 | 225 | 74 | ||
JT684042.1 | - | 2 - 3 | 230 | 74 | ||
JT695587.1 | - | 1 - 2 | 65 | 74 | ||
JT726725.1 | - | 2 - 3 | 118 | 74 | ||
JT727518.1 | - | 2 - 3 | 218 | 74 | ||
JT750471.1 | - | 2 - 3 | 229 | 74 | ||
KA337235.1 | - | 1 - 2 | 226 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
