Assay Details
Gene Symbol: MUC20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005675240.3 | XP_005675297.1 | 2 - 3 | 1656 | 71 | |
XM_018046032.1 | XP_017901521.1 | 2 - 3 | 1656 | 71 | ||
GenBank mRNA | JO428694.2 | - | 2 - 3 | 703 | 71 | |
JO430144.2 | - | 2 - 3 | 703 | 71 | ||
KA348692.1 | - | 1 - 2 | 255 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
