Assay Details
| Gene Symbol: MRPL54 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005682576.3 | XP_005682633.1 | 1 - 2 | 152 | 77 | |
| GenBank mRNA | JO610396.2 | - | 2 - 3 | 1623 | 77 | |
| JO623211.2 | - | 2 - 3 | 843 | 77 | ||
More Information
Set Membership: |
|

MRPL54
Genomic Map