Assay Details
| Gene Symbol: FAM229B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005684552.3 | XP_005684609.1 | 3 - 4 | 413 | 63 | |
| XM_005684555.3 | XP_005684612.1 | 3 - 4 | 356 | 63 | ||
| XM_005684556.3 | XP_005684613.1 | 2 - 3 | 250 | 63 | ||
| XM_018053053.1 | XP_017908542.1 | 3 - 4 | 323 | 63 | ||
| GenBank mRNA | JT736534.1 | - | 3 - 4 | 386 | 63 | |
More Information
Set Membership: |
|

FAM229B
Genomic Map