Assay Details
| Gene Symbol: SNX20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005691967.3 | XP_005692024.1 | 2 - 3 | 263 | 104 | |
| GenBank mRNA | JO577955.1 | - | 2 - 3 | 360 | 104 | |
More Information
Set Membership: |
|

SNX20
Genomic Map