Assay Details
| Gene Symbol: VPS37B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018061335.1 | XP_017916824.1 | 2 - 3 | 349 | 76 | |
| GenBank mRNA | JO427530.1 | - | 2 - 3 | 427 | 76 | |
More Information
Set Membership: |
|

VPS37B
Genomic Map