Assay Details
| Gene Symbol: TUBB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005688299.3 | XP_005688356.1 | 2 - 3 | 201 | 75 | |
| XM_018057608.1 | XP_017913097.1 | 2 - 3 | 201 | 75 | ||
| GenBank mRNA | JO338868.2 | - | 2 - 3 | 2756 | 75 | |
| JO338870.2 | - | 2 - 3 | 2753 | 75 | ||
More Information
Set Membership: |
|

TUBB1
Genomic Map