Assay Details
| Gene Symbol: TEX35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005690917.2 | XP_005690974.1 | 7 - 8 | 608 | 79 | |
| XM_018060644.1 | XP_017916133.1 | 3 - 4 | 373 | 79 | ||
| XM_018060645.1 | XP_017916134.1 | 7 - 8 | 529 | 79 | ||
More Information
Set Membership: |
|

TEX35
Genomic Map