Assay Details
| Gene Symbol: MEDAG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018056618.1 | XP_017912107.1 | 4 - 5 | 978 | 98 | |
| GenBank mRNA | JO366125.2 | - | 1 - 2 | 1439 | 98 | |
More Information
Set Membership: |
|

MEDAG
Genomic Map