Assay Details
Gene Symbol: PYCR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005694051.2 | XP_005694108.1 | 4 - 5 | 445 | 58 | |
XM_018065441.1 | XP_017920930.1 | 4 - 5 | 385 | 58 | ||
GenBank mRNA | KA306285.1 | - | 4 - 5 | 461 | 58 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
