Assay Details
| Gene Symbol: HSDL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018061779.1 | XP_017917268.1 | 3 - 4 | 777 | 56 | |
| XM_018061780.1 | XP_017917269.1 | 4 - 5 | 832 | 56 | ||
| GenBank mRNA | JO638809.2 | - | 3 - 4 | 792 | 56 | |
More Information
Set Membership: |
|

HSDL1
Genomic Map